2,3,4-Trichlorotoluene 95%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-2,3,4-trichlorobenzene, 1,2,3-Trichloro-4-methylbenzene

A115851-100MG A115851-250MG A115851-1G
103798-634EA 33 USD
103798-634 103824-496 77245-908 77245-910 77311-322
2,3,4-Trichlorotoluene 95%
2,3,4-Trichlorotoluene

Precaución: for Research Use Only

Formula: C₇H₅Cl₃
Boiling Pt: 243.5 °C (760 mm Hg)
Melting Pt: 43 °C
Flash Pt: 153.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00018098
CAS Number: 7359-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR