8-Hydroxyjulolidine 97%

Supplier: Ambeed
Warning

Synonyms: 1,2,3,5,6,7-Hexahydropyrido[3,2,1-ij]quinolin-8-ol, 2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizin-8-ol

A188329-100G A188329-100MG A188329-10G A188329-250MG A188329-1G A188329-5G A188329-25G
103816-090EA 8 USD
103816-090 103846-682 76690-764 76690-766 76690-768 76690-770 77084-410 77084-412 77302-118
8-Hydroxyjulolidine 97%
8-Hydroxyjulolidine

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO
MW: 189.26 g/mol
Boiling Pt: 374.4 °C (760 mm Hg)
Melting Pt: 132…134 °C
Flash Pt: 213.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00006918
CAS Number: 41175-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR