1,2,3,4-Tetramethylbenzene 97%

Supplier: Ambeed
Warning

A235750-1G A235750-25G A235750-5G A235750-100G A235750-10G
103811-026EA 32 USD
103811-026 103850-794 77089-806 77089-808 77089-810 77193-198 77193-200
1,2,3,4-Tetramethylbenzene 97%
1,2,3,4-Tetramethylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₄
MW: 134.22 g/mol
Boiling Pt: 203 °C (760 mm Hg)
Flash Pt: 73.88 °C
Storage Temperature: Ambient
MDL Number: MFCD00008521
CAS Number: 488-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR