1,2,3,4-Tetrahydroquinoxaline 97%

Supplier: Ambeed
Warning

A128769-100G A128769-10G A128769-1G A128769-5G
103801-180EA 7 USD
103801-180 77094-878 77094-880 77298-324 77313-530
1,2,3,4-Tetrahydroquinoxaline 97%
1,2,3,4-Tetrahydroquinoxaline

Precaución: for Research Use Only

Formula: C₈H₁₀N₂
MW: 134.18 g/mol
Melting Pt: 92…98 °C
Flash Pt: 176.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00047564
CAS Number: 3476-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR