Tetrahydroquinoline-6-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 1,2,3,4-Tetrahydroquinoline-6-carboxylic acid

A180027-5G A180027-10G A180027-250MG A180027-100MG
103807-498EA 99 USD
103807-498 103845-852 77083-330 77191-658 77191-660 77615-514
Tetrahydroquinoline-6-carboxylic acid 95%
Tetrahydroquinoline-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Boiling Pt: 382.6 °C (760 mm Hg)
Melting Pt: 168 °C
Flash Pt: 185.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00957087
CAS Number: 5382-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR