1,2,3,4-Tetrahydroquinaldine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-1,2,3,4-tetrahydroquinoline, 1,2,3,4-Tetrahydro-2-methylquinoline

A295010-1G A295010-25G A295010-5G A295010-100G
103814-038EA 13 USD
103814-038 103835-174 76837-816 76837-818 76837-820 77002-392
1,2,3,4-Tetrahydroquinaldine 98%
1,2,3,4-Tetrahydroquinaldine

Precaución: for Research Use Only

Formula: C₁₀H₁₃N
MW: 147.22 g/mol
Boiling Pt: 256.7 °C (760 mm Hg)
Flash Pt: 113.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00023886
CAS Number: 1780-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR