1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1,2,3,4-Tetrahydro-1-naphthoic acid

A103491-10G A103491-1G A103491-5G A103491-25G
103796-562EA 5 USD
103796-562 103822-268 76796-366 77283-112 77309-142 77565-788
1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid 98%
1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid

Precaución: For research use only.

Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
Melting Pt: 75…77 °C
Flash Pt: 156.408 °C
Storage Temperature: Ambient
MDL Number: MFCD02179127
CAS Number: 1914-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR