(±)-1,2,3,4-Tetrahydro-1-naphthol 97%

Supplier: Ambeed
Warning

Synonyms: ac-alpha-Tetralol, 1,2,3,4-Tetrahydronaphthalen-1-ol, 1,2,3,4-Tetrahydro-1-naphthol

A254974-250MG A254974-1G
103812-180EA 9 USD
103812-180 103852-034 76692-598 77091-494
(±)-1,2,3,4-Tetrahydro-1-naphthol 97%
(±)-1,2,3,4-Tetrahydro-1-naphthol

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Boiling Pt: 254.8 °C (760 mm Hg)
Melting Pt: 28…32 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00001739
CAS Number: 529-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR