1,2,3,4-Tetrahydro-2-naphthoic acid 97%

Supplier: Ambeed
Warning

A142906-10G A142906-1G A142906-5G
103803-222EA 95 USD
103803-222 76798-378 77299-316 77299-318
1,2,3,4-Tetrahydro-2-naphthoic acid 97%
1,2,3,4-Tetrahydro-2-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₂
MW: 176.212 g/mol
Boiling Pt: 343.9 °C (760 mm Hg)
Melting Pt: 93…96 °C
Flash Pt: 156.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00001741
CAS Number: 53440-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR