1,2,3,4-Tetrahydro-2,7-naphthyridine hydrochloride 97%

Supplier: Ambeed
Warning

A253668-1G A253668-250MG A253668-100MG
103812-102EA 216 USD
103812-102 103851-952 77304-616 77568-458 77672-268
1,2,3,4-Tetrahydro-2,7-naphthyridine hydrochloride 97%
1,2,3,4-Tetrahydro-2,7-naphthyridine hydrochloride

Precaución: For research use only.

Formula: C₈H₁₁ClN₂
Storage Temperature: Refrigerator
MDL Number: MFCD18253770
CAS Number: 1354940-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR