1,2,3,4-Tetrahydro-2,6-naphthyridine dihydrochloride 97%

Supplier: Ambeed
Warning

A119231-100MG A119231-250MG
103799-186EA 313 USD
103799-186 76870-142 76870-144
1,2,3,4-Tetrahydro-2,6-naphthyridine dihydrochloride 97%
1,2,3,4-Tetrahydro-2,6-naphthyridine dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₂Cl₂N₂
MW: 207.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11506136
CAS Number: 449175-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR