1,2,3,4-Tetrahydro-1,6-naphthyridine 97%

Supplier: Ambeed
Warning

A142752-100MG A142752-250MG
103803-208EA 73 USD
103803-208 77190-506 77190-508
1,2,3,4-Tetrahydro-1,6-naphthyridine 97%
1,2,3,4-Tetrahydro-1,6-naphthyridine

Precaución: for Research Use Only

Formula: C₈H₁₀N₂
MW: 134.178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08234763
CAS Number: 13623-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR