1,2,3,4,5-Pentamethylcyclopentadiene 95% (by GC)

Supplier: Ambeed
Danger

Synonyms: Pentamethylcyclopentadiene, 1,2,3,4,5-Pentamethylcyclopenta-1,3-diene, 1,2,3,4,5-Pentamethyl-1,3-cyclopentadiene

A608995-1G A608995-25G A608995-5G A608995-10G
103829-018EA 8 USD
103829-018 103840-156 76844-320 76844-322 76844-324 77198-098
1,2,3,4,5-Pentamethylcyclopentadiene 95% (by GC)
1,2,3,4,5-Pentamethylcyclopentadiene

Precaución: for Research Use Only

Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 170 °C at N/A mmHg
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00001354
CAS Number: 4045-44-7
UN: 3295
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR