1,2,3,3-Tetramethyl-3H-indolium iodide 98%

Supplier: Ambeed
Warning

A142082-25G A142082-100G
77316-310EA 30 USD
77316-310 77566-856
1,2,3,3-Tetramethyl-3H-indolium iodide 98%
1,2,3,3-Tetramethyl-3H-indolium iodide

Precaución: For research use only.

Formula: C₁₂H₁₆IN
Melting Pt: 258 °C
Storage Temperature: Ambient
MDL Number: MFCD00013333
CAS Number: 5418-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR