1,1'-Thiocarbonyldiimidazole 85%
Supplier: Ambeed
|
Warning
|
A107641-100G
A107641-25G
A107641-5G
103797-224EA
7
USD
103797-224
103822-990
77135-780
77135-782
77309-848
1,1'-Thiocarbonyldiimidazole 85%
1,1'-Thiocarbonyldiimidazole
Precaución: for Research Use Only
Formula:
C₇H₆N₄S Boiling Pt: 390.8 °C (760 mm Hg) Melting Pt: 98…102 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005289 CAS Number: 6160-65-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...