1,1'-Sulfonyldiimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 1,1'-sulphonyldiimidazole

A134089-100G A134089-10G A134089-25G A134089-5G A134089-500G
77162-010EA 98.88 USD
77162-010 77162-012 77314-688 77314-690 77683-190
1,1'-Sulfonyldiimidazole 98%
1,1'-Sulfonyldiimidazole

Precaución: For research use only.

Formula: C₆H₆N₄O₂S
MW: 198.20 g/mol
Melting Pt: 135…137 °C
Flash Pt: 238.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00015893
CAS Number: 7189-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR