1,1'-Ferrocenedimethanol 97%

Supplier: Ambeed
Warning

A238268-100MG A238268-250MG A238268-5G A238268-1G
103811-218EA 102 USD
103811-218 77193-270 77193-272 77193-274 77304-104
1,1'-Ferrocenedimethanol 97%
1,1'-Ferrocenedimethanol

Precaución: for Research Use Only

Formula: C₁₂H₁₄FeO₂
Melting Pt: 106…108 °C
Storage Temperature: Ambient
MDL Number: MFCD02091688
CAS Number: 1291-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR