1,1'-Ferrocenedicarboxylic acid 95%

Supplier: Ambeed
Warning

A497273-1G A497273-5G A497273-100G A497273-250MG A497273-25G
103828-264EA 8 USD
103828-264 103839-342 76843-310 76843-312 77196-966 77196-968 77307-600
1,1'-Ferrocenedicarboxylic acid 95%
1,1'-Ferrocenedicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀FeO₄
Boiling Pt: 236.7 °C (760 mm Hg)
Melting Pt: 300 °C
Flash Pt: 104.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00001423
CAS Number: 1293-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR