1,1-Dibromo-2,2-bis(chloromethyl)cyclopropane 96%
Supplier: Ambeed
|
Warning
|
A182725-1G
A182725-25G
A182725-5G
A182725-100G
A182725-10G
A182725-500G
103807-742EA
6
USD
103807-742
103846-118
77083-670
77083-672
77083-674
77191-740
77191-742
77191-744
1,1-Dibromo-2,2-bis(chloromethyl)cyclopropane 96%
1,1-Dibromo-2,2-bis(chloromethyl)cyclopropane
Precaución: for Research Use Only
Formula:
C₅H₆Br₂Cl₂ Boiling Pt: 277.1 °C (760 mm Hg) Melting Pt: 48…50 °C Flash Pt: 134.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00101445 CAS Number: 98577-44-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...