1,1-Cyclohexanediacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1,1-Cyclohexanediaceticacid

A180371-25G A180371-5G A180371-10G A180371-500G A180371-100G
103845-886EA 5 USD
103845-886 77083-372 77083-374 77191-664 77191-666 77567-656
1,1-Cyclohexanediacetic acid 97%
1,1-Cyclohexanediacetic acid

Precaución: For research use only.

Formula: C₁₀H₁₆O₄
MW: 200.23 g/mol
Boiling Pt: 405.7 °C (760 mm Hg)
Melting Pt: 67…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00001519
CAS Number: 4355-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR