1,1'-Bis(diphenylphosphino)ferrocene 97%

Supplier: Ambeed
Warning

Synonyms: DPPF, 1,1-Bis(diphenylphosphino)ferrocene

A323581-25G A323581-5G A323581-100G A323581-10G A323581-1G A323581-500G
103836-286EA 6 USD
103836-286 76839-200 76839-202 77002-894 77002-896 77002-898 77002-900
1,1'-Bis(diphenylphosphino)ferrocene 97%
1,1'-Bis(diphenylphosphino)ferrocene

Precaución: for Research Use Only

Formula: C₃₄H₂₈FeP₂
MW: 554.39 g/mol
Melting Pt: 181…182 °C
Flash Pt: 182.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00001422
CAS Number: 12150-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR