1,1'-Bis(dicyclohexylphosphino)ferrocene 97%

Supplier: Ambeed
Warning

A291164-100G A291164-10G A291164-250MG A291164-1G A291164-5G A291164-25G
103813-892EA 6 USD
103813-892 103835-034 77166-788 77166-790 77166-792 77305-594 77305-596 77683-873
1,1'-Bis(dicyclohexylphosphino)ferrocene 97%
1,1'-Bis(dicyclohexylphosphino)ferrocene

Precaución: For research use only.

Formula: C₃₄H₅₂FeP₂
MW: 570.5057 g/mol
Melting Pt: 134…136 °C
Storage Temperature: Ambient
MDL Number: MFCD09038820
CAS Number: 146960-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR