1,1-Bis(4-hydroxyphenyl)cyclohexane 98%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Cyclohexylidenebisphenol

A900823-25G A900823-5G A900823-100G A900823-500G
103841-348EA 5 USD
103841-348 76845-616 76845-618 76880-784 76880-786
1,1-Bis(4-hydroxyphenyl)cyclohexane 98%
1,1-Bis(4-hydroxyphenyl)cyclohexane

Precaución: for Research Use Only

Formula: C₁₈H₂₀O₂
MW: 268.36 g/mol
Melting Pt: 190…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00019341
CAS Number: 843-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR