1,12-Dibromododecane 98%

Supplier: Ambeed
Warning

A111960-1000G A111960-100G A111960-10G A111960-25G A111960-1G A111960-5G A111960-500G
103797-928EA 5 USD
103797-928 103823-752 77189-104 77189-106 77189-108 77189-110 77284-194 77310-560 77614-868
1,12-Dibromododecane 98%
1,12-Dibromododecane

Precaución: for Research Use Only

Formula: BrH₂C(CH₂)₁₀CH₂Br
MW: 328.13 g/mol
Boiling Pt: 280.4 °C (760 mm Hg)
Flash Pt: 123.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00000226
CAS Number: 3344-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR