Tetramethylurea 98%

Supplier: Ambeed
Warning

A727108-100G A727108-10G A727108-500G A727108-25G
103810-538EA 168 USD
103810-538 77035-938 77035-940 77035-942 77308-216
Tetramethylurea 98%
Tetramethylurea

Precaución: for Research Use Only

Formula: C₅H₁₂N₂O
Boiling Pt: 62…63 °C (12 mm Hg)
Melting Pt: 1 °C
Storage Temperature: Ambient
MDL Number: MFCD00008319
CAS Number: 632-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR