1,1,3,3-Tetramethyl-1,3-diphenyldisiloxane 97%

Supplier: Ambeed
Warning

A301500-25G A301500-5G A301500-10G
103814-308EA 6 USD
103814-308 103835-460 76838-150 76838-152 76838-154 77746-565
1,1,3,3-Tetramethyl-1,3-diphenyldisiloxane 97%
1,1,3,3-Tetramethyl-1,3-diphenyldisiloxane

Precaución: for Research Use Only

Formula: C₁₆H₂₂OSi₂
Melting Pt: …89 °C
Flash Pt: 156 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039793
CAS Number: 56-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR