1,1,2-Trimethyl-1H-benz[e]indole 98%

Supplier: Ambeed
Warning

Synonyms: 1,1,2-Trimethyl-1H-benzo[e]indole

A233168-100G A233168-10G A233168-25G A233168-5G A233168-500G
103850-632EA 8 USD
103850-632 77031-100 77031-102 77089-584 77303-956 77600-870
1,1,2-Trimethyl-1H-benz[e]indole 98%
1,1,2-Trimethyl-1H-benz[e]indole

Precaución: for Research Use Only

Formula: C₁₅H₁₅N
MW: 209.29 g/mol
Boiling Pt: 333.7 °C (760 mm Hg)
Melting Pt: 111…117 °C
Flash Pt: 147.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00082627
CAS Number: 41532-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR