N,N-Bis(trifluoromethanesulfonyl)aniline 98%

Supplier: Ambeed
Warning

Synonyms: 1,1,1-Trifluoro-N-phenyl-N-((trifluoromethyl)sulphonyl)methanesulphonamide, N-Phenylbis(trifluoromethanesulphonimide)

A108768-1000G A108768-100G A108768-10G A108768-1G A108768-250MG A108768-25G A108768-500G A108768-5G
103823-174EA 8 USD
103823-174 77093-834 77093-836 77093-838 77093-840 77093-842 77093-844 77093-846 77310-004
N,N-Bis(trifluoromethanesulfonyl)aniline 98%
N,N-Bis(trifluoromethanesulphonyl)aniline

Precaución: for Research Use Only

Formula: C₈H₅F₆NO₄S₂
MW: 357.25 g/mol
Boiling Pt: 305.3 °C (760 mm Hg)
Melting Pt: 100…102 °C
Flash Pt: 138.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00000404
CAS Number: 37595-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR