N-(2-Pyridyl)bis(trifluoromethanesulfonimide) 95%

Supplier: Ambeed
Warning

Synonyms: N,N-Bis(trifluoromethylsulphonyl)-2-pyridylamine, 2-Pyridyltriflimide, 2-[N,N-Bis(trifluoromethylsulphonyl)amino]pyridine, 1,1,1-Trifluoro-N-(pyridin-2-yl)-N-((trifluoromethyl)sulphonyl)methanesulphonamide

A113669-10G A113669-25G A113669-1G A113669-5G
103798-266EA 18 USD
103798-266 103824-108 77245-824 77245-826 77310-924 77310-926
N-(2-Pyridyl)bis(trifluoromethanesulfonimide) 95%
N-(2-Pyridyl)bis(trifluoromethanesulfonimide)

Precaución: for Research Use Only

Formula: C₇H₄F₆N₂O₄S₂
Boiling Pt: 324.8 °C (760 mm Hg)
Melting Pt: 40…42 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191834
CAS Number: 145100-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR