1H,1H,2H,2H-Perfluoro-1-iodooctane 98%

Supplier: Ambeed
Danger

Synonyms: 1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodooctane, 1H,1H,2H,2H-Perfluorooctyl iodide, 1-Iodo-3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctane

A762333-100G A762333-500G
103840-892EA 13 USD
103840-892 77747-505 77747-506
1H,1H,2H,2H-Perfluoro-1-iodooctane 98%
1H,1H,2H,2H-Perfluoro-1-iodooctane

Precaución: Keep in a dark place, inert atmosphere, 2…8 °C

Formula: C₈H₄F₁₃I
Boiling Pt: 179.2 °C (760 mm Hg)
Melting Pt: 21 °C (760 mm Hg)
Flash Pt: 80.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039410
CAS Number: 2043-57-4
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR