1,3,5-Triacetylbenzene 97%

Supplier: Ambeed
Warning

A155975-100G A155975-10G A155975-250MG A155975-1G A155975-25G A155975-5G
103804-816EA 20 USD
103804-816 103843-018 77190-942 77190-944 77190-946 77300-152 77300-154 77300-156
1,3,5-Triacetylbenzene 97%
1,3,5-Triacetylbenzene

Precaución: Sealed in dry, at room temperature.

Formula: C₁₂H₁₂O₃
MW: 204.2200 g/mol
Boiling Pt: 367.5 °C (760 mm Hg)
Melting Pt: 160…164 °C
Flash Pt: 158.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00008741
CAS Number: 779-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR