(Acetylmethylene)triphenylphosphorane 98%

Supplier: Ambeed
Warning

A259642-25G A259642-5G A259642-100G A259642-10G A259642-500G
103852-334EA 5 USD
103852-334 77091-904 77091-906 77140-684 77140-686 77140-688
(Acetylmethylene)triphenylphosphorane 98%
(Acetylmethylene)triphenylphosphorane

Precaución: for Research Use Only

Formula: C₂₁H₁₉OP
Boiling Pt: 478.5 °C (760 mm Hg)
Melting Pt: 204…209 °C
Flash Pt: 243.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00008774
CAS Number: 1439-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR