1-(Trifluoromethyl)cyclobutanecarboxylic acid 98%

Supplier: Ambeed
Warning

A103627-10G A103627-250MG A103627-1G A103627-25G
103796-582EA 59 USD
103796-582 103822-288 77107-352 77107-354 77309-162 77570-212
1-(Trifluoromethyl)cyclobutanecarboxylic acid 98%
1-(Trifluoromethyl)cyclobutanecarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₇F₃O₂
MW: 168.12 g/mol
Boiling Pt: 183 °C (760 mm Hg)
Melting Pt: 24…26 °C
Flash Pt: 64.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03425197
CAS Number: 277756-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR