4-(Trifluoromethyl)styrene 98% stabilized
Supplier: Ambeed
|
Danger
|
Synonyms:
1-(Trifluoromethyl)-4-vinylbenzene
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC)
A166538-1G
A166538-25G
A166538-5G
103806-074EA
24
USD
103806-074
103844-336
77320-590
77320-592
77320-594
4-(Trifluoromethyl)styrene 98% stabilized
4-(Trifluoromethyl)styrene
Precaución: for Research Use Only
Formula:
C₉H₇F₃ MW: 172.15 g/mol Boiling Pt: 65…66 °C (40 mm Hg) Flash Pt: 42 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00075539 CAS Number: 402-50-6 UN: 1993 ADR: 3,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...