1-(3-Tetrahydrofuryl)-1H-pyrazole-4-boronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 1-(Tetrahydrofuran-3-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1-(Oxolan-3-yl)-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

A173369-5G A173369-100MG A173369-250MG
103806-812EA 52 USD
103806-812 103845-108 77082-352 77163-630 77163-632
1-(3-Tetrahydrofuryl)-1H-pyrazole-4-boronic acid pinacol ester 95%
1-(3-Tetrahydrofuryl)-1H-pyrazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₁BN₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD18383269
CAS Number: 1029715-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR