1-(Tetrahydro-2H-pyran-4-yl)ethanone 97%

Supplier: Ambeed
Warning

Synonyms: 4-Acetyltetrahydropyran

A188041-100G A188041-10G A188041-25G A188041-1G A188041-5G
103816-066EA 5 USD
103816-066 103846-656 77029-720 77029-722 77029-724 77084-376 77084-378
1-(Tetrahydro-2H-pyran-4-yl)ethanone 97%
1-(Tetrahydro-2H-pyran-4-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 197.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08704647
CAS Number: 137052-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR