1-(Tetrahydro-2H-pyran-4-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(Tetrahydro-2H-pyran-4-yl)-1H-pyrazole-4-boronic acid Pinacol ester
A505354-1G
A505354-25G
A505354-5G
103828-322EA
10
USD
103828-322
103839-422
76843-396
76843-398
77307-630
1-(Tetrahydro-2H-pyran-4-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%
1-(Tetrahydro-2H-pyran-4-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₁₄H₂₃BN₂O₃ MW: 278.1550 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD12033229 CAS Number: 1040377-03-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...