1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 1-(Tetrahydropyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 1-(Tetrahydro-2H-pyran-2-yl)-1H-pyrazole-5-boronic acid Pinacol ester, 1-(Tetrahydropyran-2-yl)-1H-pyrazole-5-boronic acid pinacol ester, 1-(Tetrahydro-2H-pyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

A103483-100G A103483-10G A103483-25G A103483-1G A103483-5G A103483-500G
103796-560EA 6 USD
103796-560 103822-266 76970-282 76970-284 76970-286 77309-138 77309-140 77610-784
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester 97%
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₃BN₂O₃
Melting Pt: 74…78 °C
Flash Pt: 204.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09037501
CAS Number: 903550-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR