1-(2-Tetrahydropyranyl)-1H-pyrazole-4-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Tetrahydro-2H-pyran-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

A228265-100G A228265-10G A228265-250MG A228265-500G A228265-1G A228265-25G A228265-5G
103819-382EA 9 USD
103819-382 103850-290 76976-026 76976-028 76976-030 76976-032 77089-076 77089-078 77089-080
1-(2-Tetrahydropyranyl)-1H-pyrazole-4-boronic acid pinacol ester 98%
1-(2-Tetrahydropyranyl)-1H-pyrazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₃BN₂O₃
MW: 278.1550 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16556148
CAS Number: 1003846-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR