p-tert-Butylanisole 98%

Supplier: Ambeed
Warning

Synonyms: 4-tert-Butylanisole, 1-(tert-Butyl)-4-methoxybenzene

A104086-100G A104086-10G A104086-25G A104086-500G A104086-5G
103822-362EA 7 USD
103822-362 76765-342 76765-344 76765-346 76765-348 77283-194
p-tert-Butylanisole 98%
p-tert-Butylanisole

Precaución: for Research Use Only

Formula: C₁₁H₁₆O
MW: 164.25 g/mol
Boiling Pt: 80 °C (15 mm Hg)
Melting Pt: 18 °C
Flash Pt: 93.89 °C
Storage Temperature: Ambient
MDL Number: MFCD00026309
CAS Number: 5396-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR