1-tert-Butyl-4-ethynylbenzene 95%

Supplier: Ambeed
Danger

Synonyms: 1-(tert-Butyl)-4-ethynylbenzene, (4-tert-Butylphenyl)ethyne, (4-tert-Butylphenyl)acetylene, 4-tert-Butylphenylacetylene, 1-Ethynyl-4-tert-butylbenzene

A484334-10G A484334-25G A484334-5G A484334-1G
103828-160EA 8 USD
103828-160 103839-234 76774-402 76843-170 76843-172 77307-560
1-tert-Butyl-4-ethynylbenzene 95%
1-tert-Butyl-4-ethynylbenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₄
Boiling Pt: 210.9 °C (760 mm Hg)
Flash Pt: 71.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00190197
CAS Number: 772-38-3
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR