1-(tert-Butyl)-1H-pyrazol-5-amine 95%

Supplier: Ambeed
Warning

A112613-250MG A112613-1G
103798-056EA 49 USD
103798-056 103823-894 76982-814 77310-706
1-(tert-Butyl)-1H-pyrazol-5-amine 95%
1-(tert-Butyl)-1H-pyrazol-5-amine

Precaución: for Research Use Only

Formula: C₇H₁₃N₃
MW: 139.198 g/mol
Boiling Pt: 247.5±13.0 °C (760 mm Hg)
Flash Pt: 103.5 °C
Storage Temperature: Ambient
MDL Number: MFCD11054260
CAS Number: 442850-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR