1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: Pyrrolidine-3-carboxylic acid, N-BOC protected

A239798-100G A239798-10G A239798-500G A239798-25G A239798-5G A239798-1G
103811-308EA 8 USD
103811-308 103851-106 76815-370 76815-372 76815-374 77090-248 77090-250 77304-172
1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid 97%
1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₄
Boiling Pt: 337 °C (760 mm Hg)
Melting Pt: 133…138 °C
Flash Pt: 157.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179181
CAS Number: 59378-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR