1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid 96%

Supplier: Ambeed
Warning

Synonyms: 1-(tert-butoxycarbonyl)piperidine-2-carboxylic acid, 1-n-boc-2-piperidinecarboxylic acid, N-BOC-2-Piperidinecarboxylic acid

A173926-100G A173926-10G A173926-25G A173926-5G A173926-500G
103845-182EA 5 USD
103845-182 76799-478 76799-480 77082-466 77082-468 77667-418
1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid 96%
1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
Melting Pt: 130…133 °C
Storage Temperature: Ambient
MDL Number: MFCD01862877
CAS Number: 98303-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR