Indoline-2-carboxylic acid, N-BOC protected 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(tert-Butoxycarbonyl)indoline-2-carboxylic acid, 1-(tert-Butoxycarbonyl)-2,3-dihydro-1H-indole-2-carboxylic acid, 1-(tert-Butoxycarbonyl)-2-carboxyindoline
A820777-1G
A820777-25G
A820777-5G
A820777-250MG
103841-132EA
120
USD
103841-132
76845-370
76845-372
76845-374
77019-522
Indoline-2-carboxylic acid, N-BOC protected 96%
Indoline-2-carboxylic acid, N-BOC protected
Precaución: for Research Use Only
Formula:
C₁₄H₁₇NO₄ MW: 263.289 g/mol Boiling Pt: 411.6±44.0 °C (760 mm Hg) Melting Pt: 121…122 °C Flash Pt: 202.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02682411 CAS Number: 137088-51-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...