1-[(tert-Butoxy)carbonyl]-3-methylazetidine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1-(tert-Butoxycarbonyl)-3-methylazetidine-3-carboxylic acid

A603132-10G A603132-250MG
103828-988EA 18 USD
103828-988 103840-128 76878-950 76878-952
1-[(tert-Butoxy)carbonyl]-3-methylazetidine-3-carboxylic acid 97%
1-[(tert-Butoxy)carbonyl]-3-methylazetidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₄
MW: 215.246 g/mol
Boiling Pt: 320.8±35.0 °C (760 mm Hg)
Flash Pt: 147.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12406581
CAS Number: 887591-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR