1-(tert-Butoxycarbonyl)-3,3-dimethyl-4-oxopiperidine 95%

Supplier: Ambeed
Warning

Synonyms: 3,3-Dimethylpiperidin-4-one, N-BOC protected

A162085-100G A162085-10G A162085-500G A162085-1G A162085-25G
103805-572EA 12 USD
103805-572 103843-820 77191-124 77191-126 77191-128 77319-852 77319-854
1-(tert-Butoxycarbonyl)-3,3-dimethyl-4-oxopiperidine 95%
1-(tert-Butoxycarbonyl)-3,3-dimethyl-4-oxopiperidine

Precaución: Sealed in dry, at 2…8°C

Formula: C₁₂H₂₁NO₃
MW: 227.3000 g/mol
Boiling Pt: 303.4 °C (760 mm Hg)
Flash Pt: 137 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09880282
CAS Number: 324769-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR