1-(tert-Butoxycarbonyl)-3,3-difluoropiperidine-4-carboxylic acid ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 1-[(tert-butoxy)carbonyl]-3,3-difluoropiperidine-4-carboxylic acid

A332743-100MG A332743-250MG A332743-5G
103825-614EA 219 USD
103825-614 77612-906 77612-908 77683-992
1-(tert-Butoxycarbonyl)-3,3-difluoropiperidine-4-carboxylic acid ≥95%, Ambeed.Inc
1-(tert-Butoxycarbonyl)-3,3-difluoropiperidine-4-carboxylic acid

Precaución: For research use only.

Formula: C₁₁H₁₇F₂NO₄
Boiling Pt: 356.4±42.0 °C (760 mm Hg)
Flash Pt: 169.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18909780
CAS Number: 1303972-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR