1-(Quinoxalin-6-yl)ethanone 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A162199-100MG A162199-1G A162199-5G A162199-250MG
103805-588EA 137 USD
103805-588 103843-838 77011-260 77300-576 77300-578 77683-339
1-(Quinoxalin-6-yl)ethanone 98%, Ambeed.Inc
1-(Quinoxalin-6-yl)ethanone

Precaución: For research use only.

Formula: C₁₀H₈N₂O
MW: 172.18 g/mol
Boiling Pt: 318.9±22.0 °C (760 mm Hg)
Melting Pt: 106…108 °C
Flash Pt: 151.1 °C
Storage Temperature: Ambient
MDL Number: MFCD09880611
CAS Number: 83570-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR