1-(Pyrimidin-4-yl)ethanone 95%

Supplier: Ambeed
Warning

A151841-100MG A151841-250MG A151841-1G A151841-5G
103804-308EA 116 USD
103804-308 103842-458 77028-162 77028-164 77299-900 77299-902
1-(Pyrimidin-4-yl)ethanone 95%
1-(Pyrimidin-4-yl)ethanone

Precaución: for Research Use Only

Formula: C₆H₆N₂O
MW: 122.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09263821
CAS Number: 39870-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR